4-Methyl-2-hexylamine hydrochloride 95%

Supplier: Apollo Scientific
Danger

OR2894-100G OR2894-25G OR2894-5G
APOSOR2894-100GEA 69.6 EUR
APOSOR2894-100G APOSOR2894-25G APOSOR2894-5G
4-Methyl-2-hexylamine hydrochloride 95%
4-Methyl-2-hexylamine hydrochloride
Formula: C₇H₁₇N
Storage Temperature: Ambient
MDL Number: MFCD09038354
CAS Number: 13803-74-2
UN: 2925
ADR: 4.1+8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR