N-Acetyl-α-D-glucosamine

Supplier: Apollo Scientific
Warning

Synonyms: 2-Acetamido-2-deoxy-α-D-glucopyranose

OR28962-1G
APOSOR28962-1GEA 21.9 EUR
APOSOR28962-1G
N-Acetyl-α-D-glucosamine
N-Acetyl-α-D-glucosamine
Formula: C₈H₁₅NO₆
Melting Pt: 211 °C
Storage Temperature: Ambient
CAS Number: 10036-64-3
EINECS: 233-115-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR