2,5-Dimethoxythiophene 95+%

Supplier: Apollo Scientific

OR29056-1G
APOSOR29056-1GEA 74.1 EUR
APOSOR29056-1G
2,5-Dimethoxythiophene 95+%
2,5-Dimethoxythiophene
Formula: C₆H₈O₂S
MW: 144.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01096545
CAS Number: 58386-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR