6-Chloropiperonal

Supplier: Apollo Scientific
Warning

OR2927-1G OR2927-250MG
APOSOR2927-1GEA 290 EUR
APOSOR2927-1G APOSOR2927-250MG
6-Chloropiperonal
6-Chloropiperonal
Formula: C₈H₅ClO₃
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00022953
CAS Number: 15952-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR