Ethyl-2-acetylbenzenecarboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 2-acetylbenzoate

OR29770-250MG
APOSOR29770-250MGEA 87 EUR
APOSOR29770-250MG
Ethyl-2-acetylbenzenecarboxylate
Ethyl-2-acetylbenzenecarboxylate
Formula: C₁₁H₁₂O₃
MW: 192.22 g/mol
Boiling Pt: 97…99 °C/0.3mmHg
Storage Temperature: Ambient
MDL Number: MFCD00173696
CAS Number: 103935-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR