5-Bromo-2-hydroxy-3-nitropyridine 97%

Supplier: Apollo Scientific
Danger

OR29816-100G
APOSOR29816-100GEA 0 EUR
APOSOR29816-100G
5-Bromo-2-hydroxy-3-nitropyridine 97%
5-Bromo-2-hydroxy-3-nitropyridine
Formula: C₅H₃BrN₂O₃
Melting Pt: 246…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00023473
CAS Number: 15862-34-7
EINECS: 239-989-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR