1-Bromo-6-chloroisoquinoline

Supplier: Apollo Scientific
Warning

OR300111-250MG
APOSOR300111-250MGEA 58 EUR
APOSOR300111-250MG
1-Bromo-6-chloroisoquinoline
1-Bromo-6-chloroisoquinoline
Formula: C₉H₅BrClN
Storage Temperature: Ambient
MDL Number: MFCD13190244
CAS Number: 1196146-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR