5-Bromo-2-(tetrahydrofuran-3-yl)isoindolin-1-one

Supplier: Apollo Scientific
Warning

OR300220-250MG
APOSOR300220-250MGEA 493 EUR
APOSOR300220-250MG
5-Bromo-2-(tetrahydrofuran-3-yl)isoindolin-1-one
5-Bromo-2-(tetrahydrofuran-3-yl)isoindolin-1-one
Formula: C₁₂H₁₂BrNO₂
MW: 282,133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561252
CAS Number: 2227272-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR