3-Phenylpyrazin-2-amine

Supplier: Apollo Scientific
Warning

OR300239-1G OR300239-250MG
APOSOR300239-1GEA 248 EUR
APOSOR300239-1G APOSOR300239-250MG
3-Phenylpyrazin-2-amine
3-Phenylpyrazin-2-amine
Formula: C₁₀H₉N₃
MW: 171.199 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05722803
CAS Number: 41270-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR