2-Bromo-6-(4-morpholino)pyridine

Supplier: Apollo Scientific

Synonyms: 2-Bromo-6-morpholinopyridine

OR300693-5G
APOSOR300693-5GEA 0 EUR
APOSOR300693-5G
2-Bromo-6-(4-morpholino)pyridine
2-Bromo-6-(4-morpholino)pyridine
Formula: C₉H₁₁BrN₂O
MW: 243.1004 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07772824
CAS Number: 332134-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR