6-Hydroxymethylnicotinic acid ethyl ester

Supplier: Apollo Scientific
Warning

OR300696-1G OR300696-500MG OR300696-5G
APOSOR300696-1GEA 0 EUR
APOSOR300696-1G APOSOR300696-500MG APOSOR300696-5G
6-Hydroxymethylnicotinic acid ethyl ester
6-Hydroxymethylnicotinic acid ethyl ester
Formula: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD12131130
CAS Number: 35005-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR