7-Quinolinol 98%

Supplier: Apollo Scientific
Warning

Synonyms: Quinolin-7-ol, 7-Hydroxyquinoline

OR301114-5G OR301114-1G OR301114-10G
APOSOR301114-5GEA 63.8 EUR
APOSOR301114-5G APOSOR301114-1G APOSOR301114-10G
7-Quinolinol 98%
7-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 313
Melting Pt: 239…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00016730
CAS Number: 580-20-1
EINECS: 209-457-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR