4-Chloro-3-iodopyridine

Supplier: Apollo Scientific
Danger

OR301298-5G
APOSOR301298-5GEA 68.3 EUR
APOSOR301298-5G
4-Chloro-3-iodopyridine
4-Chloro-3-iodopyridine
Formula: C₅H₃ClIN
Melting Pt: 71…81 °C
Storage Temperature: Freezer
MDL Number: MFCD00234024
CAS Number: 89167-34-0
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR