1-Chloro-3-iodobenzene 98%

Supplier: Apollo Scientific
Warning

OR30192-500G
APOSOR30192-500GEA 140 EUR
APOSOR30192-500G
1-Chloro-3-iodobenzene 98%
1-Chloro-3-iodobenzene
Formula: C₆H₄ClI
Boiling Pt: 103…104 °C/15mmHg
Density: 1,926 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00001046
CAS Number: 625-99-0
EINECS: 210-920-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR