5-Amino-1-(pyridin-2-yl)-1H-pyrazole-4-carbonitrile

Supplier: Apollo Scientific

OR302058-1G
APOSOR302058-1GEA 63.8 EUR
APOSOR302058-1G
5-Amino-1-(pyridin-2-yl)-1H-pyrazole-4-carbonitrile
5-Amino-1-(pyridin-2-yl)-1H-pyrazole-4-carbonitrile
Formula: C₉H₇N₅
Storage Temperature: Ambient
MDL Number: MFCD00128357
CAS Number: 72816-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR