7-Methoxyquinoline-4-carboxylic acid

Supplier: Apollo Scientific

OR302185-1G OR302185-100MG OR302185-250MG
APOSOR302185-1GEA 748 EUR
APOSOR302185-1G APOSOR302185-100MG APOSOR302185-250MG
7-Methoxyquinoline-4-carboxylic acid
7-Methoxyquinoline-4-carboxylic acid
Formula: C₁₁H₉NO₃
MW: 203.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11108759
CAS Number: 816449-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR