6-(4-Methylpiperazin-1-yl)pyrimidin-4-amine

Supplier: Apollo Scientific

OR302506-1G
APOSOR302506-1GEA 125 EUR
APOSOR302506-1G
6-(4-Methylpiperazin-1-yl)pyrimidin-4-amine
6-(4-Methylpiperazin-1-yl)pyrimidin-4-amine
Formula: C9H15N5
MW: 193.2500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999229
CAS Number: 96225-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR