N-(1-Oxoindan-5-yl)acetamide 97%

Supplier: Apollo Scientific
Warning

OR30266-1G OR30266-25G OR30266-5G
APOSOR30266-1GEA 69.6 EUR
APOSOR30266-1G APOSOR30266-25G APOSOR30266-5G
N-(1-Oxoindan-5-yl)acetamide 97%
N-(1-Oxoindan-5-yl)acetamide
Formula: C₁₁H₁₁NO₂
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00099465
CAS Number: 58161-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR