4-Iodo-2-methoxynicotinaldehyde

Supplier: Apollo Scientific

OR303775-5G
APOSOR303775-5GEA 78.3 EUR
APOSOR303775-5G
4-Iodo-2-methoxynicotinaldehyde
4-Iodo-2-methoxynicotinaldehyde
Formula: C₇H₆INO₂
Melting Pt: 96…97 °C
Storage Temperature: Ambient
MDL Number: MFCD05662396
CAS Number: 158669-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR