Methyl-4-(pyrrolidin-1-ylmethyl)benzoate

Supplier: Apollo Scientific

OR303935-1G
APOSOR303935-1GEA 0 EUR
APOSOR303935-1G
Methyl-4-(pyrrolidin-1-ylmethyl)benzoate
Methyl-4-(pyrrolidin-1-ylmethyl)benzoate
Formula: C₁₃H₁₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD01452504
CAS Number: 160598-45-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR