N,N-Diethylazetidin-3-amine dihydrochloride

Supplier: Apollo Scientific

OR304333-10G OR304333-1G OR304333-5G
APOSOR304333-10GEA 923 EUR
APOSOR304333-10G APOSOR304333-1G APOSOR304333-5G
N,N-Diethylazetidin-3-amine dihydrochloride
N,N-Diethylazetidin-3-amine dihydrochloride
Formula: C₇H₁₆N₂·2ClH
Storage Temperature: Ambient
MDL Number: MFCD18381080
CAS Number: 149088-16-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR