3-Methyl-3,9-diazaspiro[5.5]undecane dihydrochloride

Supplier: Apollo Scientific

OR304398-1G OR304398-5G
APOSOR304398-1GEA 700 EUR
APOSOR304398-1G APOSOR304398-5G
3-Methyl-3,9-diazaspiro[5.5]undecane dihydrochloride
3-Methyl-3,9-diazaspiro[5.5]undecane dihydrochloride
Formula: C₁₀H₂₀N₂·2ClH
MW: 241.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185054
CAS Number: 13323-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR