9-Anthracenemethanol 98%

Supplier: Apollo Scientific
Warning

OR30453-100G
APOSOR30453-100GEA 65.4 EUR
APOSOR30453-100G
9-Anthracenemethanol 98%
9-Anthracenemethanol
Formula: C₁₅H₁₂O
Melting Pt: 162…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00001264
CAS Number: 1468-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR