5-Bromo-1H-indazol-3-ol

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-1,2-dihydro-3H-indazol-3-one

OR305112-1G
APOSOR305112-1GEA 84.1 EUR
APOSOR305112-1G
5-Bromo-1H-indazol-3-ol
5-Bromo-1H-indazol-3-ol
Formula: C₇H₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD07781638
CAS Number: 7364-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR