2-Bromo-3-chloroimidazo[1,2-a]pyridine

Supplier: Apollo Scientific

OR30663-1G OR30663-250MG
APOSOR30663-1GEA 250 EUR
APOSOR30663-1G APOSOR30663-250MG
2-Bromo-3-chloroimidazo[1,2-a]pyridine
2-Bromo-3-chloroimidazo[1,2-a]pyridine
Formula: C₇H₄BrClN₂
MW: 231.4800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11869753
CAS Number: 1159511-19-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR