Octachloronaphthalene 98%

Supplier: Apollo Scientific
Warning

OR30671-1G OR30671-25G OR30671-5G
APOSOR30671-1GEA 29 EUR
APOSOR30671-1G APOSOR30671-25G APOSOR30671-5G
Octachloronaphthalene 98%
Octachloronaphthalene
Formula: C₁₀Cl₈
Boiling Pt: 440 °C
Melting Pt: 194…197 °C
Density: 2 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
CAS Number: 2234-13-1
EINECS: 218-778-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR