1-[Amino(4-chlorophenyl)methyl]-2-naphthol hydrochloride

Supplier: Apollo Scientific

OR307613-1G
APOSOR307613-1GEA 590 EUR
APOSOR307613-1G
1-[Amino(4-chlorophenyl)methyl]-2-naphthol hydrochloride
1-[Amino(4-chlorophenyl)methyl]-2-naphthol hydrochloride
Formula: C₁₇H₁₅Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD07366595
CAS Number: 856209-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR