3-Amino-3-(4-isopropyl-phenyl)-propan-1-ol

Supplier: Apollo Scientific

OR307626-1G
APOSOR307626-1GEA 517 EUR
APOSOR307626-1G
3-Amino-3-(4-isopropyl-phenyl)-propan-1-ol
3-Amino-3-(4-isopropyl-phenyl)-propan-1-ol
Formula: C₁₂H₁₉NO
Storage Temperature: Ambient
MDL Number: MFCD07366613
CAS Number: 955287-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR