trans-2-(Methylamino)indan-1-ol

Supplier: Apollo Scientific

OR307671-1G
APOSOR307671-1GEA 556 EUR
APOSOR307671-1G
trans-2-(Methylamino)indan-1-ol
trans-2-(Methylamino)indan-1-ol
Formula: C₁₀H₁₃NO
MW: 163,216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11505080
CAS Number: 23671-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR