6-Bromo-4-chloroquinoline-2-carboxaldehyde

Supplier: Apollo Scientific

OR307876-1G
APOSOR307876-1GEA 459 EUR
APOSOR307876-1G
6-Bromo-4-chloroquinoline-2-carboxaldehyde
6-Bromo-4-chloroquinoline-2-carboxaldehyde
Formula: C₁₀H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD06824254
CAS Number: 904369-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR