4-Amino-6-bromoquinoline

Supplier: Apollo Scientific

OR307942-1G OR307942-5G
APOSOR307942-1GEA 119 EUR
APOSOR307942-1G APOSOR307942-5G
4-Amino-6-bromoquinoline
4-Amino-6-bromoquinoline
Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407953
CAS Number: 65340-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR