4-Amino-7-bromo-2-methylquinoline

Supplier: Apollo Scientific

OR307948-1G
APOSOR307948-1GEA 426 EUR
APOSOR307948-1G
4-Amino-7-bromo-2-methylquinoline
4-Amino-7-bromo-2-methylquinoline
Formula: C₁₀H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD09787720
CAS Number: 948293-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR