4-Hydrazino-6-methoxy-2-methylquinoline hydrochloride

Supplier: Apollo Scientific

OR307974-1G OR307974-5G
APOSOR307974-1GEA 317 EUR
APOSOR307974-1G APOSOR307974-5G
4-Hydrazino-6-methoxy-2-methylquinoline hydrochloride
4-Hydrazino-6-methoxy-2-methylquinoline hydrochloride
Formula: C₁₁H₁₃N₃O·ClH
Storage Temperature: Ambient
MDL Number: MFCD09997977
CAS Number: 1171611-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR