6,7-Dichloroquinoline-3-carboxylic acid

Supplier: Apollo Scientific

OR307995-1G
APOSOR307995-1GEA 542 EUR
APOSOR307995-1G
6,7-Dichloroquinoline-3-carboxylic acid
6,7-Dichloroquinoline-3-carboxylic acid
Formula: C₁₀H₅Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD09787797
CAS Number: 948294-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR