N-Boc-N-methyl-1,3-diaminopropane

Supplier: Apollo Scientific
Warning

OR308000-1G OR308000-5G
APOSOR308000-1GEA 49.3 EUR
APOSOR308000-1G APOSOR308000-5G
N-Boc-N-methyl-1,3-diaminopropane
N-Boc-N-methyl-1,3-diaminopropane
Formula: C₉H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01631189
CAS Number: 150349-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR