5-Amino-7-azaindole

Supplier: Apollo Scientific

Synonyms: 1H-Pyrrolo[2,3-b]pyridin-5-amine

OR308039-1G OR308039-5G
APOSOR308039-1GEA 88.6 EUR
APOSOR308039-1G APOSOR308039-5G
5-Amino-7-azaindole
5-Amino-7-azaindole
Formula: C₇H₇N₃
Storage Temperature: Ambient
MDL Number: MFCD06659683
CAS Number: 100960-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR