4-Iodo-1H-indazole

Supplier: Apollo Scientific

OR308061-1G
APOSOR308061-1GEA 122 EUR
APOSOR308061-1G
4-Iodo-1H-indazole
4-Iodo-1H-indazole
Formula: C₇H₅IN₂
Storage Temperature: Ambient
MDL Number: MFCD07781607
CAS Number: 885522-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR