6-(Hydroxymethyl)-1-methyl-1H-indazole 97%

Supplier: Apollo Scientific

OR30863-1G OR30863-250MG
APOSOR30863-1GEA 104 EUR
APOSOR30863-1G APOSOR30863-250MG
6-(Hydroxymethyl)-1-methyl-1H-indazole 97%
6-(Hydroxymethyl)-1-methyl-1H-indazole
Formula: C₉H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD11052638
CAS Number: 1092961-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR