1-(1H-Indazol-4-yl)ethanone

Supplier: Apollo Scientific
Warning

OR30884-100MG
APOSOR30884-100MGEA 119 EUR
APOSOR30884-100MG
1-(1H-Indazol-4-yl)ethanone
1-(1H-Indazol-4-yl)ethanone
Formula: C₉H₈N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11869762
CAS Number: 1159511-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR