4-(4-Bromophenyl)nicotinaldehyde

Supplier: Apollo Scientific

OR3089-1G OR3089-50MG OR3089-250MG OR3089-500MG
APOSOR3089-1GEA 470 EUR
APOSOR3089-1G APOSOR3089-50MG APOSOR3089-250MG APOSOR3089-500MG
4-(4-Bromophenyl)nicotinaldehyde
4-(4-Bromophenyl)nicotinaldehyde
Formula: C₁₂H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD01310797
CAS Number: 376646-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR