4-Bromoquinoline-3-carboxylic acid ethyl ester

Supplier: Apollo Scientific

Synonyms: Ethyl 4-bromoquinoline-3-carboxylate

OR309357-1G OR309357-250MG OR309357-5G
APOSOR309357-1GEA 89.9 EUR
APOSOR309357-1G APOSOR309357-250MG APOSOR309357-5G
4-Bromoquinoline-3-carboxylic acid ethyl ester
4-Bromoquinoline-3-carboxylic acid ethyl ester
Formula: C₁₂H₁₀BrNO₂
MW: 280.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16987479
CAS Number: 1242260-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR