(S)-Methyl 2,3-diaminopropanoate dihydrochloride

Supplier: Apollo Scientific
Danger

OR309421-1G OR309421-250MG
APOSOR309421-1GEA 71.2 EUR
APOSOR309421-1G APOSOR309421-250MG
(S)-Methyl 2,3-diaminopropanoate dihydrochloride
(S)-Methyl 2,3-diaminopropanoate dihydrochloride
Formula: C₄H₁₀N₂O₂·2ClH
MW: 191.056 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998031
CAS Number: 147857-43-0
UN: 1759
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR