2-Amino-3-chloropyridine 98%

Supplier: Apollo Scientific
Danger

OR30948-100G OR30948-25G
APOSOR30948-100GEA 95.7 EUR
APOSOR30948-100G APOSOR30948-25G
2-Amino-3-chloropyridine 98%
2-Amino-3-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD03541052
CAS Number: 39620-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR