3-(Anilinomethyl)-7-methoxyquinolin-2(1H)-one

Supplier: Apollo Scientific

OR310770-1G
APOSOR310770-1GEA 780 EUR
APOSOR310770-1G
3-(Anilinomethyl)-7-methoxyquinolin-2(1H)-one
3-(Anilinomethyl)-7-methoxyquinolin-2(1H)-one
Formula: C₁₇H₁₆N₂O₂
MW: 280.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11048615
CAS Number: 947018-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR