(4-(Bicyclo[1.1.1]pentan-1-yl)phenyl)methanamine

Supplier: Apollo Scientific

OR312402-250MG
APOSOR312402-250MGEA 1390 EUR
APOSOR312402-250MG
(4-(Bicyclo[1.1.1]pentan-1-yl)phenyl)methanamine
(4-(Bicyclo[1.1.1]pentan-1-yl)phenyl)methanamine
Formula: C₁₂H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD27996116
CAS Number: 1823872-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR