2-Methyl-3-hydroxy-6-iodopyridine 95%

Supplier: Apollo Scientific

Synonyms: 6-Iodo-2-methylpyridin-3-ol

OR314076-1G OR314076-250MG OR314076-5G
APOSOR314076-1GEA 200 EUR
APOSOR314076-1G APOSOR314076-250MG APOSOR314076-5G
2-Methyl-3-hydroxy-6-iodopyridine 95%
2-Methyl-3-hydroxy-6-iodopyridine
Formula: C₆H₆INO
Storage Temperature: Ambient
MDL Number: MFCD11044376
CAS Number: 848952-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR