3-Amino-2-methoxy-6-picoline 98%

Supplier: Apollo Scientific
Danger

OR315073-1G OR315073-5G OR315073-25G
APOSOR315073-1GEA 56.7 EUR
APOSOR315073-1G APOSOR315073-5G APOSOR315073-25G
3-Amino-2-methoxy-6-picoline 98%
3-Amino-2-methoxy-6-picoline
Formula: C₇H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD03095079
CAS Number: 186413-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR