5-Bromo-2-hydroxy-4-picoline 98%

Supplier: Apollo Scientific
Danger

OR3151-25G OR3151-5G OR3151-500MG
APOSOR3151-25GEA 200 EUR
APOSOR3151-25G APOSOR3151-5G APOSOR3151-500MG
5-Bromo-2-hydroxy-4-picoline 98%
5-Bromo-2-hydroxy-4-picoline
Formula: C₆H₆BrNO
Melting Pt: 198…202 °C
Storage Temperature: Ambient
MDL Number: MFCD03427664
CAS Number: 164513-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR