5-Ethyl-2-methylpyridine

Supplier: Apollo Scientific
Danger

OR3152-25G OR3152-100G
APOSOR3152-25GEA 21.9 EUR
APOSOR3152-25G APOSOR3152-100G
5-Ethyl-2-methylpyridine
5-Ethyl-2-methylpyridine
Formula: C₈H₁₁N
Boiling Pt: 178 °C
Melting Pt: –71 °C
Density: 0,919 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00006344
CAS Number: 104-90-5
EINECS: 203-250-0
UN: 2300
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR