3-Methyl-5-nitrobenzoic acid 95%

Supplier: Apollo Scientific

OR315323-5G
APOSOR315323-5GEA 88.6 EUR
APOSOR315323-5G
3-Methyl-5-nitrobenzoic acid 95%
3-Methyl-5-nitrobenzoic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129754
CAS Number: 113882-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR